EMAIL THIS PAGE TO A FRIEND
Journal of animal science
Analysis and pharmacokinetics of cimaterol in growing Holstein steers.
PMID 1361928
Abstract
Pharmacokinetic parameters for the beta 2-adrenergic agonist, cimaterol (CIM), were determined in growing Holstein steers. Compartmental analysis was used after measurement of CIM in body fluids by affinity chromatography and HPLC using UV detection. Recoveries from spiked plasma and urine standards were 70 +/- 1.2% and 68 +/- 1.1%, respectively. The minimum detection level in plasma was 1 ng/mL and the average CV was 5.1% for concentrations that ranged from 1 to 30 ng/mL. Four steers (276 +/- 24 kg) received 15 mg of CIM by bolus intravenous injection. Plasma CIM levels declined in a biphasic manner with half-lives of 2.5 min for the distribution phase and 54 min for the elimination phase. A two-compartment open model was used to describe the disappearance of CIM and the following pharmacokinetic parameters were obtained: central compartment volume (Vc) = .76 L/kg, apparent volume of distribution (Vd) = 4.1 L/kg, and transfer rate constants from the central to peripheral compartment (k12) = .177/min, from the peripheral to central compartment (k21) = .054/min and elimination from the central compartment (kel) = .074/min. After 8 h, total urinary CIM accounted for only 18.3% of the administered dose. Results suggest that circulating concentrations of CIM in growing steers are influenced by its accumulation in an unidentified peripheral pool and its conversion into unknown metabolite(s) before elimination.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32568
Cimaterol, VETRANAL®, analytical standard
C12H17N3O