EMAIL THIS PAGE TO A FRIEND
The Journal of antibiotics
Pneumocandins from Zalerion arboricola. V. Glutamic acid- and leucine-derived amino acids in pneumocandin A0 (L-671,329) and distinct origins of the substituted proline residues in pneumocandins A0 and B0.
PMID 1362720
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
420174
cis-3-Hydroxy-DL-proline, 95%
C5H9NO3