EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Glucocorticosteroids and transaminase activity. I. Increased activity of glutamicpyruvic transaminase in four conditions associated with gluconeogenesis.
PMID 13641244