EMAIL THIS PAGE TO A FRIEND
Eisei Shikenjo hokoku. Bulletin of National Institute of Hygienic Sciences
[Studies on "new coccine standard" for the dye standard of the National Institute of Hygienic Sciences].
PMID 1364425
Abstract
The "New Coccine Standard (C.I. 16255)" for the Dye Standard of the National Institute of Hygienic Sciences was prepared. The content of this Dye Standard determined by the titanium trichloride method was 97.5% on the average. The ultraviolet-visible absorption and infrared spectra of the Dye Standard were also measured.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
199737
New Coccine, Dye content 75 %
C20H11N2Na3O10S3
18137
Ponceau 4R, analytical standard
C20H11N2Na3O10S3