EMAIL THIS PAGE TO A FRIEND
Solid state nuclear magnetic resonance
A bi-symmetric square wave Zeeman modulator for nuclear quadrupole resonance.
PMID 1365735
Abstract
A simple circuit has been designed to generate a bi-symmetric square wave Zeeman modulation for the detection of nuclear quadrupole resonance. The square waveform not only provides an optimum result among bi-symmetric modulation waveforms, but also allows the observation of the Zeeman perturbed NQR powder pattern without the need for an extra external magnetic field.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
11618
Calcium metaborate, technical, 39-44% B2O3 basis, 31-37% CaO basis, powder
B2CaO4 · 2H2O