EMAIL THIS PAGE TO A FRIEND
Cytotechnology
Use of monoclonal antibodies in solid tumors diagnosis: the endometrial carcinoma.
PMID 1367118
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A0550
Ala-Gln, ≥98% (HPLC)
C8H15N3O4