EMAIL THIS PAGE TO A FRIEND
Journal of chemical technology and biotechnology (Oxford, Oxfordshire : 1986)
Synthesis and biological activity of some 5-substituted aminomethyl-8-hydroxyquinoline-7-sulphonic acids.
PMID 1367477
Abstract
5-Aryl (or alkyl)-8-hydroxyquinoline-7-sulphonic acids have been prepared by the Mannich reaction of 8-hydroxyquinoline-7-sulphonic acid with primary and secondary amines. Their bactericidal activities have been determined.