EMAIL THIS PAGE TO A FRIEND
Chemical communications (Cambridge, England)
Synthesis, characteristics and luminescence properties of oligo(phenylenevinylene) dimers with a biphenyl linkage center.
PMID 13678203
Abstract
Building a new structual oligo(phenylenevinylene) by linking trimeric phenylenvinylene (TPV) through the phenylphenyl bond of a central phenylene ring was reported. The resultant TPV dimers exhibit weak intermolecular interactions and intense blue photoluminescence in the solid state as well as high phase transition temperatures up to 250 degrees C. Organic light-emitting devices (OLEDs) based on these materials display blue emission with low turn-on voltage (about 3 V), maximum luminance approaching 2000 cd m(-2) and efficiency up to 1.6 cd A(-1).
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
167266
2-Bromo-1,4-dimethylbenzene, 99%
C8H9Br