EMAIL THIS PAGE TO A FRIEND
Journal of inorganic biochemistry
cis-1,4-diaminocyclohexane-Pt(II) and -(IV) adducts with DNA bases and nucleosides.
PMID 13678810
Abstract
A series of new platinum(II) and platinum(IV) adducts of type [P(II)(cis-1,4-DACH)LCl]NO(3,) where cis-1,4-DACH=cis-1,4-diaminocyclohexane, and L=9-ethylguanine, 1-methylcytosine, adenine, adenosine, cytosine, cytidine, guanine, and [Pt(IV)(cis-1,4-DACH)Ltrans-(X)(2)Cl]NO(3), (where Y=hydroxo or acetato), were synthesized and characterized by elemental analysis, infrared spectroscopy, and 1H and 195Pt nuclear magnetic resonance spectroscopy.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
32851
trans-1,4-Diaminocyclohexane, ≥98.0% (GC)
C6H14N2