EMAIL THIS PAGE TO A FRIEND
Phytochemistry
Kava lactones and the kava-kava controversy.
PMID 13679089
Abstract
Kava-kava is a traditional beverage of the South Pacific islanders and has had centuries of use without major side effects. Standardised extracts of kava-kava produced in Europe have led to many serious health problems and even to death. The extraction process (aqueous vs. acetone in the two types of preparations) is responsible for the difference in toxicity as extraction of glutathione in addition to the kava lactones is important to provide protection against hepatotoxicity. The Michael reaction between glutathione and kava lactones, resulting in opening of the lactone ring, reduces the side effects of the kava kava extracts. This protective activity was demonstrated using Acanthamoebae castellanii in which 100% cell death occurred with 100 mg ml(-1) kava lactones alone, and 40% cell death with a mixture of 100 mg ml (-1)glutathione and 100 mg ml (-1) kava lactones. A comparison of kava lactone toxicity with other pharmaceutical products is discussed and recommendations made for safe usage of kava-kava products
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
59780
DL-Kavain, analytical standard
C14H14O3