EMAIL THIS PAGE TO A FRIEND
The Journal of Clinical Investigation
Distribution patterns of extracellular matrix components and adhesion receptors are intricately modulated during first trimester cytotrophoblast differentiation along the invasive pathway, in vivo.
PMID 1370295