EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
Synthesis and substance P receptor binding activity of androstano[3,2-b]pyrimido[1,2-a]benzimidazoles.
PMID 1370695
Abstract
Several heterosteroids containing a dihydroethisterone skeleton were prepared and shown to displace substance P in a receptor binding assay. Further biochemical (kinetic and Scatchard analyses) and pharmacological evaluation (substance P-induced plasma extravasation and salivation in the rat) of a representative example in this series (5a) established that these compounds are competitive antagonists at the substance P receptor.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
W103
WIN 51708 hydrate, solid
C29H33N3O · xH2O
W104
WIN 62,577, solid
C29H31N3O