EMAIL THIS PAGE TO A FRIEND
The Biochemical journal
Haemolysins in venoms of Australian snakes. Observations on the haemolysins of the venoms of some Australian snakes and the separation of phospholipase A from the venom of Pseudechis porphyriacus.
PMID 13723433