EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the USA
CDK2 encodes a 33-kDa cyclin A-associated protein kinase and is expressed before CDC2 in the cell cycle.
PMID 1372993