EMAIL THIS PAGE TO A FRIEND
Nature
Degradation of benzylpenicillin at pH 7.5 to D-benzylpenicilloic acid.
PMID 13761466
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
68186 Benzylpenicilloic acid disodium salt, mixture of epimeres, ≥95% (HPLC)
C16H18N2Na2O5S