EMAIL THIS PAGE TO A FRIEND
Carbohydrate research
Synthesis of 1-deoxynojirimycin-containing glycans related to the Lewis X and sialyl-Lewis X epitopes recognized by LEC-CAMs.
PMID 1381282
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B5019
Benzyl 2-acetamido-2-deoxy-3-O-β-D-galactopyranosyl-α-D-galactopyranoside
C21H31NO11