EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Uronic acid metabolism in bacteria. III. Purification and properties of D-altronic acid and D-mannonic acid dehydrases in Escherichia coli.
PMID 13831814
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
12271
2-Keto-3-deoxy-D-gluconic acid lithium salt, ≥95% (TLC)
C6H10O6 · xLi+
89883
D-Altronic acid lithium salt, ≥98.5% (TLC)
C6H12O7 · xLi+