EMAIL THIS PAGE TO A FRIEND
The American journal of physiology
Effect of strophanthidin on electrolyte content and PAH accumulation of rabbit kidney slices.
PMID 13874786
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
S6626
Strophanthidin
C23H32O6