EMAIL THIS PAGE TO A FRIEND
Acta crystallographica. Section C, Crystal structure communications
Structure of calcium 2,6-difluorobenzoate dihydrate.
PMID 1388940
Abstract
Ca2+.2C7H3O2F2-.2H2O, M(r) = 390.3, monoclinic, C2/c, a = 17.584 (4), b = 10.771 (3), c = 7.887 (2) A, beta = 91.28 (2) degrees, V = 1493 A3, Z = 4, Dm = 1.75, Dx = 1.74 g cm-3, lambda(Mo K alpha) = 0.71073 A, mu = 4.92 cm-1, F(000) = 792, T = 293 K, final R = 0.037 for 2415 unique observed reflections. The eight-coordinate Ca2+ ion is linked to six carboxylate O atoms from four different 2,6-difluorobenzoate ions and two water molecules. Each 2,6-difluorobenzoate ion chelates one Ca2+ ion and forms unidentate bridging linkages to two other Ca2+ ions leading to a polymeric structure. Unlike the situation in calcium 2-fluorobenzoate, the two F atoms in this structure are not involved in significant C-F...H-O hydrogen bonding.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
190039
2,6-Difluorobenzoic acid, 98%
C7H4F2O2