EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
Investigation of serum trypsin and related substances. I. The quantitative demonstration of trypsinlike activity in human blood serum by a micromethod.
PMID 13912641