EMAIL THIS PAGE TO A FRIEND
Nature
Pyruvic acid, a unique component of an exocellular bacterial polysaccharide.
PMID 13913912
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
107360
Pyruvic acid, 98%
C3H4O3