EMAIL THIS PAGE TO A FRIEND
Biochemistry
Studies on the mechanism of action of monoamine oxidase: metabolism of N,N-dimethyltryptamine and N,N-dimethyltryptamine-N-oxide.
PMID 13914428
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
04476
Hordenine, ≥97.0% (HPLC)
C10H15NO