EMAIL THIS PAGE TO A FRIEND
Current therapeutic research, clinical and experimental
Carrageenin and cinchophen ulcer.
PMID 13945120
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
196479
2-Phenyl-4-quinolinecarboxylic acid, 99%
C16H11NO2