EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
The denaturation of proteins. IV. Conalbumin and iron(III)-conalbumin in urea solution.
PMID 13948414
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C0880 Conalbumin from chicken egg white, Iron complex