EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Experimental study of benzotrichloride metabolism].
PMID 1398177
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
147893
α,α,α-Trichlorotoluene, 98%
C7H5Cl3