EMAIL THIS PAGE TO A FRIEND
Hormone and metabolic research = Hormon- und Stoffwechselforschung = Hormones et metabolisme
Effect of the aquaretic vasopressin antagonists d(CH2)5[D-Tyr(Et)2-Val4]AVP and d(CH2)5[D-Phe2-Phe4]AVP on urine production in healthy dogs.
PMID 1398466