EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
Polygalacturonic acid metabolism in bacteria. II. Formation and metabolism of 3-deoxy-D-glycero-2, 5-hexodiulosonic acid.
PMID 13986017
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
89883
D-Altronic acid lithium salt, ≥98.5% (TLC)
C6H12O7 · xLi+