EMAIL THIS PAGE TO A FRIEND
Biochemical pharmacology
Comparison of some biologgical and biochemical properties of 6-bromopurine and 6-iodopurine.
PMID 13986581
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
104981
6-Bromopurine, 98%
C5H3BrN4