EMAIL THIS PAGE TO A FRIEND
Nederlands tijdschrift voor geneeskunde
[Severe psychological side effects in children using high-dosage deptropine].
PMID 1407169
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
D0406200 Deptropine citrate, European Pharmacopoeia (EP) Reference Standard
C23H27NO · C6H8O7