EMAIL THIS PAGE TO A FRIEND
Medicina experimentalis : International journal of experimental medicine
[ON THE EFFECT OF DRUGS ON THE CLEARANCE OF I-131 LABELED ROSE BENGAL BY THE RAT LIVER].
PMID 14086235
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
196479
2-Phenyl-4-quinolinecarboxylic acid, 99%
C16H11NO2