EMAIL THIS PAGE TO A FRIEND
Proceedings of the National Academy of Sciences of the United States of America
Defective guanyl nucleotide-binding protein beta gamma subunits in a forskolin-resistant mutant of the Y1 adrenocortical cell line.
PMID 1409589
Abstract
Forskolin-resistant mutants derived from Y1 adrenocortical cells display decreased responsiveness both to receptor and postreceptor stimulators of adenylyl cyclase and decreased amounts of the alpha subunits of the GTP-binding proteins (G proteins) that mediate stimulation (Gs) and inhibition (Gi) of adenylyl cyclase--namely, Gs alpha and Gi alpha-2. This phenotype is suggestive of a mutation that affects the processing or plasma membrane incorporation of G protein alpha subunits. Since the membrane attachment of heterotrimeric G proteins has been ascribed in part to the beta gamma subunits, we examined the quantity and functional activity of beta gamma subunits in wild-type Y1 and forskolin-resistant Forsk-10r-9 and Forsk-10r-3 cells. We now show that two assays previously used to examine the activity of purified beta gamma subunits--namely, to support either rhodopsin-catalyzed guanyl nucleotide exchange on Gt alpha or pertussis toxin-catalyzed ADP-ribosylation of Gt alpha--can be used with detergent extracts of cells. In both assays the beta gamma activity in Forsk-10r-9 and Forsk-10r-3 extracts was decreased by 53-76% compared with wild-type Y1 extracts. When normalized for immunoreactive beta subunit, the beta gamma activity in the Forsk-10r-9 samples was decreased by 55-57% compared with the wild-type Y1 samples. These results suggest that a mutation of one of the G protein beta or gamma subunits may result in the multiple defects of adenylyl cyclase activity and apparent loss of G protein alpha subunits seen in the forskolin-resistant mutant cells. The frequency with which these spontaneous mutations arise in the Y1 cell line suggests that they may contribute more generally to genetic abnormalities in signal transduction.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
details
Y1, 85062807