EMAIL THIS PAGE TO A FRIEND
Biochimica et biophysica acta
SPECTRAL ABSORPTION PROPERTIES OF ORDINARY AND FULLY DEUTERIATED CHLOROPHYLLS A AND B.
PMID 14104939
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
C5753
Chlorophyll a, from spinach
C55H72MgN4O5
C5878
Chlorophyll b from spinach, ≥90% (HPLC), ≤0.5% Chlorophyll a
C55H70MgN4O6