EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
NUCLEOSIDE TRIPHOSPHATE-NUCLEOSIDE DIPHOSPHATE TRANSPHOSPHORYLASE (NUCLEOSIDE DIPHOSPHOKINASE). I. ISOLATION OF THE CRYSTALLINE ENZYME FROM BREWERS' YEAST.
PMID 14114857