EMAIL THIS PAGE TO A FRIEND
Diseases of the nervous system
THE ADRENOCHROME THEORY OF SCHIZOPHRENIA: A REVIEW.
PMID 14130059
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A5752
Adrenochrome
C9H9NO3