EMAIL THIS PAGE TO A FRIEND
Journal of immunology (Baltimore, Md. : 1950)
STUDIES ON HEAT-LABILE OPSONIN IN RABBIT SERUM.
PMID 14139537
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
R5131 Anti-Rabbit Serum antibody produced in goat, whole antiserum