EMAIL THIS PAGE TO A FRIEND
Agents and actions
Separate induction of human blood platelet aggregation or cytotoxicity by different concentrations of PAF-acether and thrombin.
PMID 1414687
Abstract
Using decreasing concentrations of PAF-acether or thrombin, it was possible to observe on human platelets, first, aggregation, classically associated to activation, then , below a threshold, cytotoxicity towards Schistosoma mansoni larvae, proposed here as stimulation. These two activities appeared as distinct and antithetic. However, their induction might be the consequence of triggering of the same receptors with different intensity, since PAF-induced, but not thrombin-induced, cytotoxicity could be inhibited with specific PAF-antagonists BN 52021 and BN 52024 also known to inhibit PAF-induced aggregation. These results give credit to the hypothesis that haemostatic and cytotoxic properties of platelets are two distinct functions of these blood elements.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
89556
Ginkgolide J, analytical standard
C20H24O10