EMAIL THIS PAGE TO A FRIEND
Analytical biochemistry
An ultraviolet-spectrophotometric method with 2-cyanoacetamide for the determination of the enzymatic degradation of reducing polysaccharides.
PMID 1416030
Abstract
A rapid, facile, and sensitive uv-spectrophotometric assay has been developed for the determination of the enzymatic degradation of polysaccharides that generates reducing sugars. The assay was carried out with 2-cyanoacetamide in a single test tube. The solution was left at pH 9 by the addition of borate buffer within 5 min. Measurement of the reaction mixture at 274 nm allows a simple determination up to 600 mumol/liter of reducing sugars. The coefficient of variation was less than 2% on all measurements. The assay was developed with pectin and polygalacturonic acid from apples and has been compared with the Somogyi-Nelson method. The new assay was then exemplarily used for the determination of the enzymatic hydrolysis products of pectin from cotton.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
B4059
Barium hydroxide solution, 0.3 N
H2BaO2
108448
Cyanoacetamide, 99%
C3H4N2O