EMAIL THIS PAGE TO A FRIEND
Archives of biochemistry and biophysics
NITRILASE. I. OCCURRENCE, PREPARATION, AND GENERAL PROPERTIES OF THE ENZYME.
PMID 14165487
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
04529 Nitrilase, recombinant, expressed in E. coli, ≥2.0 U/mg