EMAIL THIS PAGE TO A FRIEND
Journal of medicinal chemistry
SUBSTITUTED 2,3-DIHYDRO-1,5-BENZOTHIAZEPIN-4(5H)-ONES AND 3,4-DIHYDRO-2-PHENYL-(2H)-1,6-BENZOTHIAZOCIN-5(6H)-ONES.
PMID 14173579
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
541850
2-Amino-4-chlorobenzenethiol, 96%
C6H6ClNS