EMAIL THIS PAGE TO A FRIEND
Bioorganicheskaia khimiia
[Synthesis of oligodeoxyribonucleotides containing 5-fluoro-2'-deoxyribocytidine].
PMID 1417995
Abstract
A synthesis of the phosphoroamidite derivative of 5-fluoro-2'-deoxycytidine which allows one to introduce the modified nucleoside residue into the 5'-position of the oligodeoxyribonucleotide by the standard solid phase phosphoroamidite method, has been carried out. Oligonucleotides with 5-fluoro-2'-deoxycytidine residues in various positions of the DNA strand are obtained by the combination of chemical and enzymatic syntheses.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
F5307
5−Fluoro−2′−deoxycytidine, ≥98% (HPLC), powder
C9H12FN3O4