EMAIL THIS PAGE TO A FRIEND
Virology
AGAR SUSPENSION CULTURE FOR THE SELECTIVE ASSAY OF CELLS TRANSFORMED BY POLYOMA VIRUS.
PMID 14187925
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
A4675 Agar, Type E, suitable for plant cell culture