EMAIL THIS PAGE TO A FRIEND
Minerva medica
[ON THE TREATMENT OF GOUT. LESSONS ON THE COURSE OF THERAPY. II. PHARMACOLOGICAL TREATMENT].
PMID 14199454
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
196479
2-Phenyl-4-quinolinecarboxylic acid, 99%
C16H11NO2