EMAIL THIS PAGE TO A FRIEND
Journal of pharmaceutical and biomedical analysis
Isolation and characterization of the S-glucuronide of 4-nitrothiophenol formed by microsomal glucuronyl transferase.
PMID 1420469
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
N27209
4-Nitrothiophenol, technical grade, 80%
C6H5NO2S