EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
METABOLISM AND FUNCTION OF BACTERIAL LIPIDS. II. BIOSYNTHESIS OF PHOSPHOLIPIDS IN ESCHERICHIA COLI.
PMID 14213340
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
90756
CDP-ethanolamine sodium salt hydrate, ≥93.0% (HPLC)
C11H20N4O11P2 · xNa+ · yH2O