EMAIL THIS PAGE TO A FRIEND
Journal of lipid research
PREPARATION OF FATTY ACID METHYL ESTERS AND DIMETHYLACETALS FROM LIPIDS WITH BORON FLUORIDE--METHANOL.
PMID 14221106
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
33356 BF3 - Methanol, 10 % (w/w), pkg of 20 × 1 mL