EMAIL THIS PAGE TO A FRIEND
Carcinogenesis
Peroxidative activation of o-phenylhydroquinone leads to the formation of DNA adducts in HL-60 cells.
PMID 1423859
Abstract
Using 32P-postlabeling we studied DNA adduct formation in HL-60 cells treated with the o-phenylphenol metabolites o-phenylhydroquinone (o-PHQ) and o-phenylbenzoquinone (o-PBQ). Treatment with 25-500 microM o-PHQ for 8 h produced one principal and three minor adducts with a relative distribution of 80, 10, 6 and 4%. The relative adduct levels from these treatments were 0.26-2.31 adducts/10(7) nucleotides. Treatment with 25-250 microM o-PBQ for 2 h resulted in a similar level of DNA modification and adduct distribution. Reaction of purified calf thymus DNA with o-PBQ produced one DNA adduct, which did not correspond to the major adduct produced in HL-60 cells. These results show that o-PHQ and o-PBQ can form DNA adducts. Peroxidase activation of o-phenylphenol may therefore play a role in the carcinogenic effect of this compound.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
227811
2-Phenylhydroquinone, 97%
C12H10O2