EMAIL THIS PAGE TO A FRIEND
The Journal of biological chemistry
THE STEREOSPECIFIC CONVERSION OF STEARIC ACID TO OLEIC ACID.
PMID 14253467
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
O1008
Oleic acid, ≥99% (GC)
C18H34O2