EMAIL THIS PAGE TO A FRIEND
Gigiena i sanitariia
[Toxicokinetics of borates and borides after a single exposure].
PMID 1427239
Abstract
We're sorry this abstract is currently not available.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
553913
Magnesium boride, powder, −100 mesh, ≥99% trace metals basis
B2Mg