EMAIL THIS PAGE TO A FRIEND
Journal of the American Chemical Society
A HIGHLY REACTIVE COLORED REAGENT WITH SELECTIVITY FOR THE TRYPTOPHAN RESIDUE IN PROTEINS. 2-HYDROXY-5-NITROBENZYL BROMIDE.
PMID 14284628