EMAIL THIS PAGE TO A FRIEND
International journal of peptide and protein research
Apparent specific volumes of some dipeptides (containing L-valine and L-leucine in aqueous alkylurea solutions).
PMID 1428531
Abstract
The apparent molal volumes of nine dipeptides containing glycine, L-valine, and L-leucine have been determined in methyl, N,N'-dimethyl and ethylurea solutions from precise density measurements. Limiting partial molal volumes. V2(0), at various solute concentrations have also been calculated. The experimental values of V2(0) in water agree reasonably well with those calculated as the sum of V2(0) of both acids after accounting for the electrostrictive effect and loss of water. There is no correlation between the values of V2(0) of individual dipeptides in alkylureas which means that the intrinsic volume and the electrostrictive effect make the largest contribution to V2(0). The contribution from other effects is within the limit of experimental error. The volumes of transfer from water to alkylurea solutions are all positive and reflect by and large the electrostrictive effect.
Related Materials
Product #
Image
Description
Molecular Formula
Add to Cart
261394
1,1-Dimethylurea, 99%
C3H8N2O
E51007
N-Ethylurea, 97%
C3H8N2O